Tisk…

Diosmin

Dodavatel: Apollo Scientific

Synonyms: 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

BIK9004-100G BIK9004-25G
APOSBIK9004-100GEA 1990 CZK
APOSBIK9004-100G APOSBIK9004-25G
Diosmin
Diosmin
Vzorec: C₂₈H₃₂O₁₅
MW: 608,5 g/mol
Bod tání: 274 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009772
CAS číslo: 520-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR