α-Ketoglutaric acid monosodium salt
Dodavatel: Apollo Scientific
|
Varování
|
BIK8011-5G
BIK8011-25G
BIK8011-50G
APOSBIK8011-5GEA
1330
CZK
APOSBIK8011-5G
APOSBIK8011-25G
APOSBIK8011-50G
α-Ketoglutaric acid monosodium salt
α-Ketoglutaric acid monosodium salt
Vzorec:
C₅H₅NaO₅ MW: 168,08 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00064196 CAS číslo: 22202-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...