Tisk…

Kanamycin sulphate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

BIK0126-100G BIK0126-25G BIK0126-250G
APOSBIK0126-100GEA 4370 CZK
APOSBIK0126-100G APOSBIK0126-25G APOSBIK0126-250G
Kanamycin sulphate
Kanamycin sulphate

Kanamycin is an aminoglycoside antibiotic and isused as a selective agent for the incorporation of the NPT II (APH3') gene in plant tissue


  • approx. 800 micrograms per mg
  • Cell culture tested

Vzorec: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS číslo: 25389-94-0
EINECS: 246-933-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR