Imipenem monohydrate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(5R,6S)-3-((2-Formimidamidoethyl)thio)-6-((R)-1-hydroxyethyl)-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate
BII5001-100MG
BII5001-10MG
BII5001-1G
APOSBII5001-100MGEA
1580
CZK
APOSBII5001-100MG
APOSBII5001-10MG
APOSBII5001-1G
Imipenem monohydrate
Imipenem monohydrate
Vzorec:
C₁₂H₁₇N₃O₄S MW: 317.36 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09753321 CAS číslo: 74431-23-5 EINECS: 264-734-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...