Bolton-Hunter reagent (sulpho-SHPP), technická kvalita
Dodavatel: Apollo Scientific
|
Varování
|
BII110-1G
BII110-5G
APOSBII110-1GEA
2280
CZK
APOSBII110-1G
APOSBII110-5G
Bolton-Hunter reagent (sulpho-SHPP), technická kvalita
Bolton-Hunter reagent (sulpho-SHPP)
Vzorec:
C₁₃H₁₃NO₅ Bod tání: 132…133 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005515 CAS číslo: 34071-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...