Tisk…

Inosine

Dodavatel: Apollo Scientific

Synonyms: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

BII1001-5G BII1001-10G BII1001-100G
APOSBII1001-5GEA 622 CZK
APOSBII1001-5G APOSBII1001-10G APOSBII1001-100G
Inosine
Inosine
Vzorec: C₁₀H₁₂N₄O₅
MW: 268,23 g/mol
Bod tání: 212…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00066770
CAS číslo: 58-63-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR