Tisk…

Ionomycin calcium salt

Dodavatel: Apollo Scientific
Varování

BII0123-10MG BII0123-5MG
APOSBII0123-10MGEA 8070 CZK
APOSBII0123-10MG APOSBII0123-5MG
Ionomycin calcium salt
Ionomycin calcium salt

Potent and selective calcium ionophore.

Vzorec: C₄₁H₇₀CaO₉
Bod tání: 204…206 °C
Storage Temperature: Freezer
MDL Number: MFCD00151183
CAS číslo: 56092-82-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR