Hematoxylin
Dodavatel: Apollo Scientific
|
Varování
|
APOSBIH6002-50GEA
6850
CZK
APOSBIH6002-50G
APOSBIH6002-10G
APOSBIH6002-25G
Hematoxylin
Hematoxylin
Vzorec:
C₁₆H₁₄O₆ MW: 302,28 g/mol Bod tání: 200 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS číslo: 517-28-2 EINECS: 208-237-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...