Tisk…

bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)

Dodavatel: Apollo Scientific
Varování

BIH1003-100MG BIH1003-1G BIH1003-500MG
APOSBIH1003-100MGEA 5200 CZK
APOSBIH1003-100MG APOSBIH1003-1G APOSBIH1003-500MG
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)

Fluorescent DNA stain-17/03/09 SR+RB

Vzorec: C₂₅H₂₄N₆O·3HCl·xH₂O
Bod tání: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD00012679
CAS číslo: 23491-45-4
EINECS: 245-690-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR