D-Glucose-6-phosphate dipotassium salt hydrate
Dodavatel: Apollo Scientific
|
Varování
|
BIG5009-1G
BIG5009-5G
BIG5009-500MG
APOSBIG5009-1GEA
4570
CZK
APOSBIG5009-1G
APOSBIG5009-5G
APOSBIG5009-500MG
D-Glucose-6-phosphate dipotassium salt hydrate
D-Glucose-6-phosphate dipotassium salt hydrate
Vzorec:
C₆H₁₁K₂O₉P Storage Temperature: Freezer |
CAS číslo:
5996-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...