Tisk…

D-(-)-Glucamine

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Amino-1-deoxy-D-glucitol, D-Glucamine, 1-Amino-1-deoxy-D-sorbitol, (-)-1-Amino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-Aminohexane-1,2,3,4,5-pentaol

BIG4884-5G BIG4884-25G
APOSBIG4884-5GEA 2900 CZK
APOSBIG4884-5G APOSBIG4884-25G
D-(-)-Glucamine
D-(-)-Glucamine
Vzorec: H₂NCH₂(CH(OH))₄CH₂OH
MW: 181.19
Bod varu: 531
Storage Temperature: Ambient
MDL Number: MFCD00077776
CAS číslo: 488-43-7
EINECS: 207-677-3
Merck Index: 12,04457

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR