Tisk…

D-(+)-Glucurono-6,3-lactone ≥99%

Dodavatel: Apollo Scientific
Varování

Synonyms: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone

BIG1110-100G BIG1110-1KG BIG1110-500G
APOSBIG1110-100GEA 622 CZK
APOSBIG1110-100G APOSBIG1110-1KG APOSBIG1110-500G
D-(+)-Glucurono-6,3-lactone ≥99%
D-(+)-Glucurono-6,3-lactone
Vzorec: C₆H₈O₆
Hustota: 0,67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135622
CAS číslo: 32449-92-6
EINECS: 251-053-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR