Tisk…

Thiazovivin ≥99%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Benzyl-2-(pyrimidin-4-ylamino)thiazole-4-carboxamide

BIFK0055-10MG BIFK0055-1MG BIFK0055-5MG
APOSBIFK0055-10MGEA 4570 CZK
APOSBIFK0055-10MG APOSBIFK0055-1MG APOSBIFK0055-5MG
Thiazovivin ≥99%
Thiazovivin

Thiazovivin is a Rho Kinase (ROCK) inhibitor that inhibits the apoptosis of human embryonic stem cells (hESCs) upon dissociation.


  • Free base
  • Purity>99%.

Vzorec: C₁₅H₁₃N₅OS
MW: 311,36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16495823
CAS číslo: 1226056-71-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR