Purvalanol A
Dodavatel: Apollo Scientific
|
Varování
|
BIFK0054-10MG
BIFK0054-5MG
APOSBIFK0054-10MGEA
1330
CZK
APOSBIFK0054-10MG
APOSBIFK0054-5MG
Purvalanol A
Purvalanol A
Free base. Purity>99%. Purvalanol A is a cell-permeable, potent, and selective cyclin-dependent kinase (CDK) inhibitor.
Vzorec:
C₁₉H₂₅N₆OCl Storage Temperature: Freezer |
MDL Number:
MFCD02179211 CAS číslo: 212844-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...