Vincristine sulphate
Dodavatel: Apollo Scientific
BIFK0048-100MG
BIFK0048-10MG
BIFK0048-50MG
APOSBIFK0048-100MGEA
11180
CZK
APOSBIFK0048-100MG
APOSBIFK0048-10MG
APOSBIFK0048-50MG
Vincristine sulphate
Vincristine sulphate
Vincristine is a mitotic inhibitor.
Vzorec:
C₄₆H₅₈N₄O₁₄S MW: 923.04 g/mol Bod tání: 300 °C Storage Temperature: Freezer |
MDL Number:
MFCD00084729 CAS číslo: 2068-78-2 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...