Tipifarnib free base ≥98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(R)-6-(Amino(4-chlorophenyl)(1-methyl-1H-imidazol-5-yl)methyl)-4-(3-chlorophenyl)-1-methylquinolin-2(1H)-one
BIFK0043-10MG
BIFK0043-5MG
BIFK0043-2MG
APOSBIFK0043-10MGEA
8070
CZK
APOSBIFK0043-10MG
APOSBIFK0043-5MG
APOSBIFK0043-2MG
Tipifarnib free base ≥98%
Tipifarnib
Vzorec:
C₂₇H₂₂Cl₂N₄O MW: 489.3958 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD07772347 CAS číslo: 192185-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...