PIK75 ≥99%
Dodavatel: Apollo Scientific
Synonyms:
N'-((6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulphonohydrazide hydrochloride, PIK 75, (E)-N'-((6-bromoH-imidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulphonohydrazide hydrochloride, PIK-75 HCl, PIK75 Hydrochloride, PIK 75 Hydrochloride
BIFK0038-25MG
BIFK0038-50MG
APOSBIFK0038-25MGEA
1620
CZK
APOSBIFK0038-25MG
APOSBIFK0038-50MG
PIK75 ≥99%
PIK75
Vzorec:
C₁₆H₁₅BrClN₅O₄S MW: 488,74 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12546130 CAS číslo: 372196-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...