Tisk…

Ruxolitinib phosphate ≥99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (R)-3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanenitrile phosphate

BIFK0028-10MG BIFK0028-25MG BIFK0028-50MG
APOSBIFK0028-10MGEA 1870 CZK
APOSBIFK0028-10MG APOSBIFK0028-25MG APOSBIFK0028-50MG
Ruxolitinib phosphate ≥99%
Ruxolitinib phosphate

Phosphate Salt, >99%

Vzorec: C₁₇H₂₁N₆O₄P
MW: 404,37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18452860
CAS číslo: 1092939-17-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR