Tisk…

Imatinib mesylate ≥99%

Dodavatel: Apollo Scientific

BIFK0026-1G BIFK0026-5G BIFK0026-250MG
APOSBIFK0026-1GEA 3310 CZK
APOSBIFK0026-1G APOSBIFK0026-5G APOSBIFK0026-250MG
Imatinib mesylate ≥99%
Imatinib mesylate

Imatinib is the active ingredient in the drug sold under the trade name Gleevec® in USA or Glivec in Europe and Australia.

Vzorec: C₂₉H₃₁N₇O·CH₄SO₃
Bod tání: 203…224 °C
Storage Temperature: Freezer
MDL Number: MFCD04307699
CAS číslo: 220127-57-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR