Tisk…

Imatinib ≥99%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide

BIFK0025-10G BIFK0025-1G BIFK0025-5G
APOSBIFK0025-10GEA 7050 CZK
APOSBIFK0025-10G APOSBIFK0025-1G APOSBIFK0025-5G
Imatinib ≥99%
Imatinib

Imatinib is the active ingredient in the drug sold under the trade name Gleevec® in USA or Glivec in Europe and Australia.

Vzorec: C₂₉H₃₁N₇O
Bod tání: 209…221 °C
Storage Temperature: Freezer
MDL Number: MFCD05662257
CAS číslo: 152459-95-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR