Tisk…

Gemcitabine free base ≥99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

BIFK0023-100MG BIFK0023-50MG BIFK0023-250MG
APOSBIFK0023-100MGEA 2070 CZK
APOSBIFK0023-100MG APOSBIFK0023-50MG APOSBIFK0023-250MG
Gemcitabine free base ≥99%
Gemcitabine
Vzorec: C₉H₁₁F₂N₃O₄
Bod tání: 203…219 °C
Storage Temperature: Freezer
MDL Number: MFCD00869720
CAS číslo: 95058-81-4
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR