Tisk…

Epirubicin hydrochloride

Dodavatel: Apollo Scientific

Synonyms: (8S,10S)-10-(((2R,4S,5R,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride, Epirubicin HCl

BIFK0018-10MG BIFK0018-5MG BIFK0018-50MG
APOSBIFK0018-10MGEA 3310 CZK
APOSBIFK0018-10MG APOSBIFK0018-5MG APOSBIFK0018-50MG
Epirubicin hydrochloride
Epirubicin hydrochloride
Vzorec: C₂₇H₂₉NO₁₁·HCl
MW: 579,98 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00941448
CAS číslo: 56390-09-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR