Tisk…

CUDC-101 free base ≥99%

Dodavatel: Apollo Scientific

BIFK0015-10MG BIFK0015-5MG BIFK0015-1MG
APOSBIFK0015-10MGEA 0 CZK
APOSBIFK0015-10MG APOSBIFK0015-5MG APOSBIFK0015-1MG
CUDC-101 free base ≥99%
CUDC-101
Vzorec: C₂₄H₂₆N₄O₄
Storage Temperature: Freezer
MDL Number: MFCD15528940
CAS číslo: 1012054-59-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR