Tisk…

Cediranib free base ≥99%

Dodavatel: Apollo Scientific

Synonyms: AZD2171, 4-((4-Fluoro-2-methyl-1H-indol-5-yl)oxy)-6-methoxy-7-(3-(pyrrolidin-1-yl)propoxy)quinazoline

BIFK0013-10MG BIFK0013-5MG BIFK0013-1MG
APOSBIFK0013-10MGEA 4370 CZK
APOSBIFK0013-10MG APOSBIFK0013-5MG APOSBIFK0013-1MG
Cediranib free base ≥99%
Cediranib
Vzorec: C₂₅H₂₇FN₄O₃
Storage Temperature: Freezer
MDL Number: MFCD09954115
CAS číslo: 288383-20-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR