Rolipram ≥99%
Dodavatel: Apollo Scientific
|
Varování
|
BIFK0003-100MG
BIFK0003-250MG
APOSBIFK0003-100MGEA
3540
CZK
APOSBIFK0003-100MG
APOSBIFK0003-250MG
Rolipram ≥99%
Rolipram
Rolipram has two enantiomeric forms, R(-) and S(+). This product is the racemic mixture of the two enantiomers.
Vzorec:
C₁₆H₂₁NO₃ Storage Temperature: Freezer |
MDL Number:
MFCD00270906 CAS číslo: 61413-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...