Riboflavine-5'-phosphate sodium salt USP
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Vitamin B2 phosphate sodium salt, Riboflavin 5'-Monophosphate sodium salt, Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogen phosphate, Flavin mononucleotide, Lactoflavin phosphate sodium salt, Riboflavin phosphate sodium
BIF4012-100G
BIF4012-25G
BIF4012-1KG
APOSBIF4012-100GEA
14490
CZK
APOSBIF4012-100G
APOSBIF4012-25G
APOSBIF4012-1KG
Riboflavine-5'-phosphate sodium salt USP
Riboflavine-5'-phosphate sodium salt
Vzorec:
C₁₇H₂₀N₄NaO₉P Storage Temperature: Freezer |
MDL Number:
MFCD00065362 CAS číslo: 130-40-5 EINECS: 204-988-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...