Tisk…

Fumonisin B1 (from Fusarium moniliforme)

Dodavatel: Apollo Scientific
Varování

BIF1004-1MG BIF1004-5MG
APOSBIF1004-1MGEA 7670 CZK
APOSBIF1004-1MG APOSBIF1004-5MG
Fumonisin B1 (from Fusarium moniliforme)
Fumonisin B1
Vzorec: C₃₄H₅₉NO₁₅
Storage Temperature: Freezer
MDL Number: MFCD00133349
CAS číslo: 116355-83-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR