Tisk…

Flavone

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Phenylchromen-4-one

BIF1002-25G BIF1002-5G
APOSBIF1002-25GEA 10150 CZK
APOSBIF1002-25G APOSBIF1002-5G
Flavone
Flavone
Vzorec: C₁₅H₁₀O₂
MW: 222.24
Bod tání: 94…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006825
CAS číslo: 525-82-6
EINECS: 208-383-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR