Tisk…

Ellagic acid

Dodavatel: Apollo Scientific
Varování

BIE1001-5G BIE1001-25G BIE1001-100G
APOSBIE1001-5GEA 744 CZK
APOSBIE1001-5G APOSBIE1001-25G APOSBIE1001-100G
Ellagic acid
Ellagic acid

Glutathione S-transference inhibitor.

Vzorec: C₁₄H₆O₈
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006914
CAS číslo: 476-66-4
EINECS: 207-508-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR