Tisk…

Ethylenediaminetetraacetic acid diammonium salt hydrate

Dodavatel: Apollo Scientific
Varování

Synonyms: EDTA diammonium salt hydrate

BIE0512-25G BIE0512-100G BIE0512-500G
APOSBIE0512-25GEA 953 CZK
APOSBIE0512-25G APOSBIE0512-100G APOSBIE0512-500G
Ethylenediaminetetraacetic acid diammonium salt hydrate
Ethylenediaminetetraacetic acid diammonium salt hydrate
Vzorec: C₁₀H₂₂N₄O₈·2H₂O
MW: 326.31 g/mol (anhy)
Bod tání: 218 °C
Storage Temperature: Ambient
MDL Number: MFCD00150463
CAS číslo: 20824-56-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR