Tisk…

N,N-Dimethyl-1-dodecylamine N-oxide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N-ethyl-N-oxido-dodecan-1-amine, LDAO, Lauryldimethylamine oxide, N,N-Dimethyldodecan-1-amine oxide, Lauramine oxide, N,N-Dimethyl-N-dodecylamine N-oxide

BID6003-5G BID6003-10G BID6003-25G
APOSBID6003-5GEA 4570 CZK
APOSBID6003-5G APOSBID6003-10G APOSBID6003-25G
N,N-Dimethyl-1-dodecylamine N-oxide
N,N-Dimethyl-1-dodecylamine N-oxide
Vzorec: C₁₄H₃₁NO
Storage Temperature: Refrigerator
MDL Number: MFCD00002049
CAS číslo: 1643-20-5
UN: 3259
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR