Daidzin
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
BID5420-100MG
BID5420-250MG
BID5420-500MG
APOSBID5420-100MGEA
4570
CZK
APOSBID5420-100MG
APOSBID5420-250MG
APOSBID5420-500MG
Daidzin
Daidzin
Vzorec:
C₂₁H₂₀O₉ MW: 416,38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00017466 CAS číslo: 552-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...