Tisk…

(3,4-Dihydroxyphenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

BID2105-100G
APOSBID2105-100GEA 4040 CZK
APOSBID2105-100G
(3,4-Dihydroxyphenyl)acetic acid 98%
(3,4-Dihydroxyphenyl)acetic acid
Vzorec: (C₆H₃(OH)₂)CH₂COOH
MW: 168,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004338
CAS číslo: 102-32-9
EINECS: 203-024-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR