Tisk…

17-DMAG hydrochloride 98%

Dodavatel: Apollo Scientific

Synonyms: 17-Dimethylaminoethylamino-17-demethoxygeldanamycin hydrochloride

BID1020-5MG BID1020-10MG BID1020-25MG
APOSBID1020-5MGEA 2150 CZK
APOSBID1020-5MG APOSBID1020-10MG APOSBID1020-25MG
17-DMAG hydrochloride 98%
17-DMAG hydrochloride
Vzorec: C₃₂H₄₈N₄O₈·HCl
MW: 653,21 g/mol
Storage Temperature: Freezer
CAS číslo: 467214-21-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR