Tisk…

2'-Deoxyadenosine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

BID1006-10G BID1006-25G BID1006-50G
APOSBID1006-10GEA 870 CZK
APOSBID1006-10G APOSBID1006-25G APOSBID1006-50G
2'-Deoxyadenosine
2'-Deoxyadenosine
Vzorec: C₁₀H₁₃N₅O₃
MW: 251,24 g/mol
Bod tání: 187…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056003
CAS číslo: 958-09-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR