Tisk…

Dicamba

Dodavatel: Apollo Scientific
Varování

BID0159-1G BID0159-25G BID0159-5G
APOSBID0159-1GEA 622 CZK
APOSBID0159-1G APOSBID0159-25G APOSBID0159-5G
Dicamba
Dicamba

  • Plant tissue culture tested

Vzorec: C₈H₆Cl₂O₃
Bod tání: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00055283
CAS číslo: 1918-00-9
EINECS: 217-635-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR