Trimethyl-β-cyclodextrin
Dodavatel: Apollo Scientific
Synonyms:
Heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin, Trimethyl-β-cyclodextrin
BICY1077-5G
APOSBICY1077-5GEA
10910
CZK
APOSBICY1077-5G
Trimethyl-β-cyclodextrin
Trimethyl-beta-cyclodextrin
Vzorec:
C₆₃H₁₁₂O₃₅ MW: 1429,55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010728 CAS číslo: 55216-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...