Tisk…
Methyl-β-cyclodextrin (mixture of several Methylated) (DS~12)
Dodavatel: Apollo Scientific
Synonyms: Methylated-β-Cyclodextrin, Methyl-β-cyclodextrin
BICY1002-25G BICY1002-5G BICY1002-100G
APOSBICY1002-25GEA 7670 CZK
APOSBICY1002-25G APOSBICY1002-5G APOSBICY1002-100G
Methyl-β-cyclodextrin (mixture of several Methylated) (DS~12)
Methyl-beta-cyclodextrin (mixture of several Methylated)
MW: 1303.3000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074980
CAS číslo: 128446-36-6