4-Methoxyphenyl 4,6-O-Benzylidene-beta-D-galactopyranoside 98%
Dodavatel: Apollo Scientific
BICL5067-1G
BICL5067-2G
BICL5067-5G
APOSBICL5067-1GEA
11180
CZK
APOSBICL5067-1G
APOSBICL5067-2G
APOSBICL5067-5G
4-Methoxyphenyl 4,6-O-Benzylidene-beta-D-galactopyranoside 98%
4-Methoxyphenyl 4,6-O-Benzylidene-beta-D-galactopyranoside
Vzorec:
C₂₀H₂₂O₇ MW: 374,39 g/mol Storage Temperature: Ambient |
CAS číslo:
176299-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...