2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside 98%
Dodavatel: Apollo Scientific
BICL5013-1G
BICL5013-5G
APOSBICL5013-1GEA
817
CZK
APOSBICL5013-1G
APOSBICL5013-5G
2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside 98%
2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside
Vzorec:
C₁₆H₁₈ClN₅O₇ Storage Temperature: Ambient |
MDL Number:
MFCD03425473 CAS číslo: 16321-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...