Tisk…

N-Methyldeoxynojirimycin 99% min

Dodavatel: Apollo Scientific

Synonyms: N-Methylmoranoline, (2R,3R,4R,5S)-2-(Hydroxymethyl)-1-methylpiperidine-3,4,5-triol

BICL4350-10MG BICL4350-25MG BICL4350-5MG
APOSBICL4350-10MGEA 10750 CZK
APOSBICL4350-10MG APOSBICL4350-25MG APOSBICL4350-5MG
N-Methyldeoxynojirimycin 99% min
N-Methyldeoxynojirimycin
Vzorec: C₇H₁₅NO₄
MW: 177,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133609
CAS číslo: 69567-10-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR