Tisk…

N-Acetylneuraminic acid methyl ester 97% min

Dodavatel: Apollo Scientific

Synonyms: Methyl 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosonate, (2S,4S,5R,6R)-Methyl 5-acetamido-2,4-dihydroxy-6-((1R,2R)-1,2,3-trihydroxypropyl)tetrahydro-2H-pyran-2-carboxylate

BICL4322-1G BICL4322-5G
APOSBICL4322-1GEA 1580 CZK
APOSBICL4322-1G APOSBICL4322-5G
N-Acetylneuraminic acid methyl ester 97% min
N-Acetylneuraminic acid methyl ester
Vzorec: C₁₂H₂₁NO₉
MW: 323,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276447
CAS číslo: 22900-11-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR