3-Deoxy-D-glucosone 95% min
Dodavatel: Apollo Scientific
|
Varování
|
BICL4267-10MG
BICL4267-5MG
BICL4267-1MG
APOSBICL4267-10MGEA
16340
CZK
APOSBICL4267-10MG
APOSBICL4267-5MG
APOSBICL4267-1MG
3-Deoxy-D-glucosone 95% min
3-Deoxy-D-glucosone
Vzorec:
C₆H₁₀O₅ MW: 162.141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01723101 CAS číslo: 4084-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...