1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose 98% min
Dodavatel: Apollo Scientific
BICL4254-1G
BICL4254-2G
BICL4254-5G
APOSBICL4254-1GEA
8010
CZK
APOSBICL4254-1G
APOSBICL4254-2G
APOSBICL4254-5G
1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose 98% min
1,2,3,4-Tetra-O-acetyl-alpha-L-fucopyranose
Vzorec:
C₁₄H₂₀O₉ Storage Temperature: Ambient |
MDL Number:
MFCD00069791 CAS číslo: 64913-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...