Tisk…

1,6-Anhydro-2,3,4-tri-O-benzyl-beta-D-glucopyranose 98% min

Dodavatel: Apollo Scientific

Synonyms: 2,3,4-Tri-O-benzyl-1,6-anhydro-beta-D-glucopyranose

BICL4209-10G BICL4209-2G BICL4209-5G
APOSBICL4209-10GEA 31840 CZK
APOSBICL4209-10G APOSBICL4209-2G APOSBICL4209-5G
1,6-Anhydro-2,3,4-tri-O-benzyl-beta-D-glucopyranose 98% min
1,6-Anhydro-2,3,4-tri-O-benzyl-beta-D-glucopyranose
Vzorec: C₂₇H₂₈O₅
Storage Temperature: Ambient
MDL Number: MFCD02683260
CAS číslo: 10548-46-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR