Tisk…

Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside 99% min

Dodavatel: Apollo Scientific

BICL4169-10G BICL4169-25G BICL4169-50G
APOSBICL4169-10GEA 9990 CZK
APOSBICL4169-10G APOSBICL4169-25G APOSBICL4169-50G
Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside 99% min
Methyl-2,3,6-tri-O-benzoyl-alpha-D-galactopyranoside
Vzorec: C₂₈H₂₆O₉
Storage Temperature: Ambient
MDL Number: MFCD06200842
CAS číslo: 3601-36-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR