Tisk…

4-Methoxyphenyl-β-D-glucopyranoside 99% min

Dodavatel: Apollo Scientific

BICL4136-5G BICL4136-1G BICL4136-2G
APOSBICL4136-5GEA 17920 CZK
APOSBICL4136-5G APOSBICL4136-1G APOSBICL4136-2G
4-Methoxyphenyl-β-D-glucopyranoside 99% min
4-Methoxyphenyl-beta-D-glucopyranoside
Vzorec: C₁₃H₁₈O₇
Storage Temperature: Ambient
MDL Number: MFCD06797143
CAS číslo: 6032-32-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR