Tisk…

L(+)-Citrulline 99+%

Dodavatel: Apollo Scientific
Varování

Synonyms: H-Cit-OH

BIC1342-25G BIC1342-100G
APOSBIC1342-25GEA 622 CZK
APOSBIC1342-25G APOSBIC1342-100G
L(+)-Citrulline 99+%
L(+)-Citrulline
Vzorec: C₆H₁₃N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00064397
CAS číslo: 372-75-8
EINECS: 206-759-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR