Cytochalasin C
Dodavatel: Apollo Scientific
|
Nebezpečí
|
BIC1015-1MG
BIC1015-5MG
APOSBIC1015-1MGEA
6850
CZK
APOSBIC1015-1MG
APOSBIC1015-5MG
Cytochalasin C
Cytochalasin C
Vzorec:
C₃₀H₃₇NO₆ MW: 507,63 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00066080 CAS číslo: 22144-76-9 UN: 1544 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...