Tisk…

Citrinin

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Citrinin, yellow, (3S,4R)-8-hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid

BIC1010-10MG BIC1010-5MG
APOSBIC1010-10MGEA 12830 CZK
APOSBIC1010-10MG APOSBIC1010-5MG
Citrinin
Citrinin
Vzorec: C₁₃H₁₄O₅
MW: 250,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006912
CAS číslo: 518-75-2
EINECS: 208-257-2
UN: 3462
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR